4-Fluoro-3,5-dimethylbenzyl bromide 98%
Leverandør: Apollo Scientific
|
Danger
|
PC303089-10G
PC303089-5G
PC303089-1G
PC303089-25G
APOSPC303089-10GEA
2875
NOK
APOSPC303089-10G
APOSPC303089-5G
APOSPC303089-1G
APOSPC303089-25G
4-Fluoro-3,5-dimethylbenzyl bromide 98%
4-Fluoro-3,5-dimethylbenzyl bromide
Formel:
C₉H₁₀BrF MW: 217,08 g/mol Smeltepunkt: 44…46 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD06660268 CAS-nummer: 886501-82-2 FN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...