[6-(Trifluoromethyl)pyridin-3-yl]methylamine 96%

Leverandør: Apollo Scientific
Warning

Synonymer: 3-(Aminomethyl)-6-(trifluoromethyl)pyridine

PC3773-1G PC3773-5G PC3773-25G
APOSPC3773-1GEA 610 NOK
APOSPC3773-1G APOSPC3773-5G APOSPC3773-25G
[6-(Trifluoromethyl)pyridin-3-yl]methylamine 96%
[6-(Trifluoromethyl)pyridin-3-yl]methylamine
Formel: C₇H₇F₃N₂·HCl
MW: 176,14 g/mol
Kokepunkt: 62…64 °C/0,4mmHg
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD01862649
CAS-nummer: 387350-39-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR