Methyl amino(4-fluorophenyl)acetate hydrochloride 98%
Leverandør: Apollo Scientific
Warning
Synonymer: 4-Fluoro-DL-phenylglycine methyl ester hydrochloride, Methyl 2-amino-2-(4-fluorophenyl)acetate hydrochloride
PC50089-1G PC50089-5G PC50089-10G PC50089-25G
APOSPC50089-1GEA 450 NOK
APOSPC50089-1G APOSPC50089-5G APOSPC50089-10G APOSPC50089-25G
Methyl amino(4-fluorophenyl)acetate hydrochloride 98%
Methyl amino(4-fluorophenyl)acetate hydrochloride
Formel: C₉H₁₁ClFNO₂
MW: 219,64 g/mol
Smeltepunkt: 177…179 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09152734
CAS-nummer: 42718-18-3