3-Bromo-4-fluorophenyl isothiocyanate
Leverandør: Apollo Scientific
Danger
PC503009-1G PC503009-25G PC503009-5G
APOSPC503009-1GEA 1075 NOK
APOSPC503009-1G APOSPC503009-25G APOSPC503009-5G
3-Bromo-4-fluorophenyl isothiocyanate
3-Bromo-4-fluorophenyl isothiocyanate
Formel: C₇H₃BrFNS
MW: 232,08 g/mol
Lagringstemperatur: Romtemperatur
CAS-nummer: 710351-24-9