3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride 98%
Leverandør: Apollo Scientific
Warning
Synonymer: 3-(Trifluoromethyl)-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride, 3-(Trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride, 3-Trifluoromethyl-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
PC51048-100G PC51048-25G
APOSPC51048-100GEA 2150 NOK
APOSPC51048-100G APOSPC51048-25G
3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride 98%
3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
Formel: C6H8ClF3N4
MW: 228,60 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09817638
CAS-nummer: 762240-92-6