1-Bromo-2-chloro-5-fluoro-4-nitrobenzene 98%

Leverandør: Apollo Scientific
Warning

PC53455-1G PC53455-5G
APOSPC53455-1GEA 1500 NOK
APOSPC53455-1G APOSPC53455-5G
1-Bromo-2-chloro-5-fluoro-4-nitrobenzene 98%
1-Bromo-2-chloro-5-fluoro-4-nitrobenzene
Formel: C₆H₂BrClFNO₂
MW: 254,4400 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD11110551
CAS-nummer: 1027833-17-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR