1-Amino-2,2,2-trifluoretan 99%

Leverandør: Apollo Scientific
Danger

Synonymer: 2,2,2-Trifluoroethylamine

PC7320-100G PC7320-25G PC7320-5G
APOSPC7320-100GEA 0 NOK
APOSPC7320-100G APOSPC7320-25G APOSPC7320-5G
1-Amino-2,2,2-trifluoretan 99%
1-Amino-2,2,2-trifluoretan
Formel: C₂H₄F₃N
MW: 99,06 g/mol
Kokepunkt: 36…38 °C
Tetthet: 1,259 g/cm³ (25 °C)
Flash punkt: –16 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00008132
CAS-nummer: 753-90-2
EINECS: 212-041-3
FN: 3286
ADR: 3+6.1+8,II

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR