Chlorodimethyl-3,3,3-trifluoropropylsilane 97%

Leverandør: Apollo Scientific
Danger

PC7820F-50G PC7820F-100G
APOSPC7820F-50GEA 2075 NOK
APOSPC7820F-50G APOSPC7820F-100G
Chlorodimethyl-3,3,3-trifluoropropylsilane 97%
Chlorodimethyl-3,3,3-trifluoropropylsilane
Formel: C₅H₁₀ClF₃Si
MW: 190,67 g/mol
Kokepunkt: 118 °C
Tetthet: 1,117 g/cm³ (25 °C)
Flash punkt: 20 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00053211
CAS-nummer: 1481-41-0
EINECS: 216-039-3
FN: 2985
ADR: 3+8,II

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR