4-(1,1,2,2-Tetrafluoroethoxy)benzoic acid 97%

Leverandør: Apollo Scientific
Warning

PC8721-100G PC8721-25G PC8721-5G
APOSPC8721-100GEA 5625 NOK
APOSPC8721-100G APOSPC8721-25G APOSPC8721-5G
4-(1,1,2,2-Tetrafluoroethoxy)benzoic acid 97%
4-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
Formel: C₉H₆F₄O₃
Smeltepunkt: 176…177 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00155926
CAS-nummer: 10009-25-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR