Ethyl-2-fluoro-5-methylbenzoate 95%

Leverandør: Apollo Scientific

Synonymer: Ethyl 2-fluoro-5-methylbenzoate

PC903903-5G
APOSPC903903-5GEA 1125 NOK
APOSPC903903-5G
Ethyl-2-fluoro-5-methylbenzoate 95%
Ethyl-2-fluoro-5-methylbenzoate
Formel: C₁₀H₁₁FO₂
MW: 182,1916 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06204763
CAS-nummer: 496841-90-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR