2-{[3-(Trifluoromethyl)benzoyl]amino}acetic acid 97%

Leverandør: Apollo Scientific
Warning

Synonymer: 2-(3-(Trifluoromethyl)benzamido)acetic acid

PC9489-1G PC9489-250MG
APOSPC9489-1GEA 1075 NOK
APOSPC9489-1G APOSPC9489-250MG
2-{[3-(Trifluoromethyl)benzoyl]amino}acetic acid 97%
2-{[3-(Trifluoromethyl)benzoyl]amino}acetic acid
Formel: C₁₀H₈F₃NO₃
MW: 247.18 g/mol
Smeltepunkt: 134…138 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00029782
CAS-nummer: 17794-48-8

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR