Stopcocks, with GL screw thread
Leverandør: Bohlender
E686-14 E688-25 E684-18 E684-14 E684-25 E688-14 E686-18 E688-18 E686-25
229-0084EA 2175 NOK
229-0084 201-1712 229-0058 229-0057 229-0059 229-0086 229-0085 229-0087 229-0107
Stopcocks, with GL screw thread
Haner
PTFE.
  • Excellent chemical resistance, the fluid medium only comes into contact with PTFE
  • Temperature resistant from −30 to +150 °C
  • Pressure resistant up to a maximum of 6 bar, vacuum suitable
Two-way stopcocks with straight bore and two threaded connectors or three-way stopcocks with L-shaped or T-shaped bore with three threaded connectors. Cylindrical stopcock for good leakproof properties, stop valve with flow direction marking. To distribute liquids or gases. Fast and simple interruption of the flow. Connection of hoses or tubes using GL laboratory screw connections.