Syringe filters, cellulose nitrate, Puradisc™, Whatman™
Leverandør: Whatman products (Cytiva)
10462000 10462500 10462510 10462520
514-1116EA 4025 NOK
514-1116 514-1126 514-0101 514-0490
Syringe filters, cellulose nitrate, Puradisc™, Whatman™
Filter Sprøytefilter
Whatman Puradisc syringe filters from Cytiva's business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 ml.
  • From the Puradisc FP 30 range
  • Eight different material options
  • Choice of 4 mm, 13 mm, and 30 mm syringe filter diameter to suit sample volume
  • Range of pore sizes for retaining a variety of particles
  • Optional sterile, medical-grade blister pack
  • Optional tube-tip format for accurate dispensing into a micro-vial
Puradisc syringe filters cater to a wide variety ofMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Dimensjoner 26×34 mm
Maks. bemanningstrykk 6,9 bar
Vekt 4,7 g