ESD safety ankle boots, Lace-Up, Lena GTX Mid
Leverandør: ELTEN
746131/34 746131/35 746131/36 746131/37 746131/38 746131/39 746131/40 746131/41 746131/42
ELTN746131/34EA 1175 NOK
ELTN746131/34 ELTN746131/35 ELTN746131/36 ELTN746131/37 ELTN746131/38 ELTN746131/39 ELTN746131/40 ELTN746131/41 ELTN746131/42
ESD safety ankle boots, Lace-Up, Lena GTX Mid
Sko og støvler
Ladies' safety ankle boot is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • GORE-TEX Performance Comfort footwear
  • Full, padded bellows tongue
  • PU/PU sole, TREKKING LADY
  • PU toe protection
  • Steel toe cap
Cowhide leather construction. Full-length inlay sole, LADIES ESD. Non-metallic, penetration-resistant midsole.
Sertifiseringer: EN ISO 20345 S3 CI/SRC, form B.