Grade 0860 ½ qualitative filter papers, Whatman™
Leverandør: Whatman products (Cytiva)
10334547 10334551 10334553 10334607
512-0524EA 1175 NOK
512-0524 512-0525 512-0526 WHAT10334607
Grade 0860 ½ qualitative filter papers, Whatman™
Filter Filterpapir Kvalitativt filterpapir
Comparable to Grade 0858 but with a smooth surface, slightly thinner and faster.
  • Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
  • Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
  • Increased total loading capacity as more filter area is available
  • Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself
More Product Information
Bestill nå
SPESIFIKASJONER
Nr. 0860 ½
Environmentally Preferable
Vekt 75 g/m²
Tykkelse 170 µm
Partikkelretensjon 7 - 12 µm
Filtreringshastighet 60 s (Herzberg)