VWR® Maximum Protection, Cleanroom Sleeve Protectors, Sterile
Leverandør: VWR Collection
113-9997EA 6400 NOK
113-9997 113-9998
VWR® Maximum Protection, Cleanroom Sleeve Protectors, Sterile
Engangsplagg
VWR® Maximum Protection sterile sleeves are manufactured from a specially formulated breathable microporous fabric that provides significant fluid and barrier protection. Garments exhibit excellent water vapour transmission to optimise user comfort.
  • Excellent breathability and water vapour transmission rate
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding
  • Durable yet soft, cloth-like fabric
  • Gamma irradiated to SAL of 10⁻⁶
Sleeves feature elastic openings and sonically welded seams and are designed with thumb loops.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Kjønn
Antrekksapplikasjoner Cleanroom
Particle shedding (Helmke Drum) Level 1
Tykkelse 0,21 mm
Vekt 56 g/m²