Biotin-PEG4-acid 98 Titrimetric

Leverandør: TCI

Synonymer: Biotin-PEG4-propionic acid, 15-(Biotinamido)-4,7,10,13-tetraoxapentadecanoic Acid, PEG4-biotin acid, 17-Oxo-21-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-4,7,10,13-tetraoxa-16-azahenicosanoic acid

B5562-250MG B5562-50MG
TCIAB5562-250MGEA 3575 NOK
TCIAB5562-250MG TCIAB5562-50MG
Biotin-PEG4-acid 98 Titrimetric
Biotin-PEG4-acid
Formel: C₂₁H₃₇N₃O₈S
MW: 491.60 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD08457831
CAS-nummer: 721431-18-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR