CloneDetect
Leverandør: MOLECULAR DEVICES
K8205
MLDVK8205EA
0
NOK
MLDVK8205
CloneDetect
Assay
Cellulær analyse
Screen and select high value mammalian clones with label-free, in situ detection of IgG secretion with the ClonePix system, and other cell analysers with a green fluorescent channel.
- Screen and select high-value mammalian clones with label-free, in situ detection of IgG secretion
- A choice of detection agents specific for secreted mouse, rat or human IgGs
- Sterile, virus and mycoplasma free
- Optimised for ClonePix system, and compatible with cell analysers with a green fluorescent channel
A cell-culture validated, sterile and azide-free antibody reagent for detecting IgG in mammalian cell clone screening.