General purpose upright freezers, TSG series
Leverandør: Thermo Scientific
TSG700FESBV TSG700FESCV TSG1500FESCV TSG400FESBV TSG400FESCV TSG1500FESBV
471-1331EA 47220 NOK
471-1331 471-1333 471-1322 471-1326 471-1328 471-1320
General purpose upright freezers, TSG series
Frysere Frysere, generelt bruk
Designed for everyday laboratory needs the Thermo Scientific™ TSG series general purpose laboratory freezers are designed with your needs in mind. The TSG series range of freezers feature the performance, security, and quality required for your laboratory applications.
  • Freezers: −20 °C set point, settable range −25 to −10 °C
  • Wide range of sizes from solid door 400 to 1500 L double door
  • Whisper-quiet operation of <60 dBA
  • Adaptive defrost
  • White painted interior and exterior
  • Temperature, door ajar, and power failure (with battery backup) alarms
  • Low heat
More Product Information
Limited Time Sale
Select items below are currently on sale
Bestill nå
SPESIFIKASJONER
Modell TSG700FESBV TSG700FESCV TSG1500FESCV TSG400FESBV TSG400FESCV TSG1500FESBV
Environmentally Preferable
Avriming Adaptive defrost Adaptive defrost Adaptive defrost Adaptive defrost Adaptive defrost Adaptive defrost
Elektronisk 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz
Hyller/skuffer 4 shelves 4 shelves 8 shelves 5 shelves 5 shelves 8 shelves
Dørtype 1 solid door 1 solid door 2 solid doors 1 solid door 1 solid door 2 solid doors
Støpsel EU EU EU EU EU EU