Rosiglitazone Related Compounds, Reference Standard
Leverandør: USP
1605817 1605828
1605817.EA 4175 NOK
1605817. 1605828.
Rosiglitazone Related Compounds, Reference Standard
Standarder Organiske standarder
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Sertifiseringer: US Pharmacopea Reference Standard
Bestill nå
SPESIFIKASJONER
Beskrivelse Rosiglitazone Maleate Rosiglitazone Related Compound A
Synonym(s) (5Z)-5-{[4-({2-[methyl(2-pyridinyl)amino]ethyl}oxy)phenyl]methylidene}-1,3-thiazolidine-2,4-dione
CAS-nr 122320-73-4 160596-25-8