Norfloxacin Related Compounds, Reference Standard
Leverandør: USP
1471550 1471583 1471619 1471517
1471550.EA 15100 NOK
1471550. 1471583. 1471619. 1471517.
Norfloxacin Related Compounds, Reference Standard
Standarder Organiske standarder
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Sertifiseringer: US Pharmacopea Reference Standard
Bestill nå
SPESIFIKASJONER
Beskrivelse Norfloxacin Related Compound E Norfloxacin Related Compound H Norfloxacin Related Compound K Norfloxacin Related Compound A
Synonym(s) 7-Chloro-1-ethyl-4-oxo-6-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid 7-[4-(ethoxycarbonyl)piperazin-1-yl]-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 6-Fluoro-1-methyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid 7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
CAS-nr 75001-78-4 105440-01-5 70459-07-3 68077-26-9
Formel C₁₆H₁₈ClN₃O₃ C₁₉H₂₂FN₃O₅ C₁₅H₁₆FN₃O₃