Multi-Purpose Tear Resistant Tags for M610, M611 and M710 (with media adapter)
Leverandør: Brady
174185 174289
BRDY174185EA 955 NOK
BRDY174185 BRDY174289
Multi-Purpose Tear Resistant Tags for M610, M611 and M710 (with media adapter)
Etiketter
Durable Non-Adhesive Polyethylene (B-109) is a multipurpose, matte finish tag material that can be used for a variety of tag applications.
  • Extremely tear resistant and able to withstand cold temperatures
  • Ideal material for both thermal transfer and dot matrix printing methods
  • Service temperature of −40 to +49 °C (−40 to +120 °F)
  • Matte finish
Applications include data and telecommunications labelling, wire and cable labelling, inventory and inspection labelling, lockout tagout, and safety identification.
Brady authentic materials work seamlesslyMore Product Information