ISAspher HPLC Columns, Diol, HILIC
Leverandør: ISERA
A111-G23A00 A111-G23B00 A111-G23C00 A111-G23D00 A111-G23E00 A111-G23F00 A111-G23G00 A111-G23H00 A111-G23J00 A111-G23K00 A111-G23L00 A111-G23M00 A111-G23N00 A111-G23P00 A111-G23Q00 A111-G23R00 A111-G23S00 A111-G23T00 A111-G23U00 A111-G23V00 A111-G23W00 A111-G23X00 A111-G23Y00 A111-G23Z00 A111-G25A00 A111-G25B00 A111-G25C00 A111-G25D00 A111-G25E00 A111-G25F00 A111-G25G00 A111-G25H00 A111-G25J00 A111-G25K00 A111-G25L00 A111-G25M00 A111-G25N00 A111-G25P00 A111-G25Q00 A111-G25R00 A111-G25S00 A111-G25T00 A111-G25U00 A111-G25V00 A111-G25W00 A111-G25X00 A111-G25Y00 A111-G25Z00 A111-G29A00 A111-G29B00 A111-G29C00 A111-G29D00 A111-G29E00 A111-G29F00 A111-G29G00 A111-G29H00 A111-G29J00 A111-G29K00 A111-G29L00 A111-G29M00 A111-G29N00 A111-G29P00 A111-G29Q00 A111-G29R00 A111-G29S00 A111-G29T00 A111-G29U00 A111-G29V00 A111-G29W00 A111-G29X00 A111-G29Y00 A111-G29Z00 A112-G25A30 A112-G25B30 A112-G25C30 A112-G25E30 A112-G25F30 A112-G25G30 A112-G25A00 A112-G25B00 A112-G25C00 A112-G25E00 A112-G25F00 A112-G25G00 A112-G23A30 A112-G23B30 A112-G23C30 A112-G23E30 A112-G23F30 A112-G23G30 A112-G23A00 A112-G23B00 A112-G23C00 A112-G23E00 A112-G23F00 A112-G23G00 A112-G29A30 A112-G29B30 A112-G29C30 A112-G29E30 A112-G29F30 A112-G29G30 A112-G29A00 A112-G29B00 A112-G29C00 A112-G29E00 A112-G29F00 A112-G29G00
ISERA111-G23A00EA 4075 NOK
ISERA111-G23A00 ISERA111-G23B00 ISERA111-G23C00 ISERA111-G23D00 ISERA111-G23E00 ISERA111-G23F00 ISERA111-G23G00 ISERA111-G23H00 ISERA111-G23J00 ISERA111-G23K00 ISERA111-G23L00 ISERA111-G23M00 ISERA111-G23N00 ISERA111-G23P00 ISERA111-G23Q00 ISERA111-G23R00 ISERA111-G23S00 ISERA111-G23T00 ISERA111-G23U00 ISERA111-G23V00 ISERA111-G23W00 ISERA111-G23X00 ISERA111-G23Y00 ISERA111-G23Z00 ISERA111-G25A00 ISERA111-G25B00 ISERA111-G25C00 ISERA111-G25D00 ISERA111-G25E00 ISERA111-G25F00 ISERA111-G25G00 ISERA111-G25H00 ISERA111-G25J00 ISERA111-G25K00 ISERA111-G25L00 ISERA111-G25M00 ISERA111-G25N00 ISERA111-G25P00 ISERA111-G25Q00 ISERA111-G25R00 ISERA111-G25S00 ISERA111-G25T00 ISERA111-G25U00 ISERA111-G25V00 ISERA111-G25W00 ISERA111-G25X00 ISERA111-G25Y00 ISERA111-G25Z00 ISERA111-G29A00 ISERA111-G29B00 ISERA111-G29C00 ISERA111-G29D00 ISERA111-G29E00 ISERA111-G29F00 ISERA111-G29G00 ISERA111-G29H00 ISERA111-G29J00 ISERA111-G29K00 ISERA111-G29L00 ISERA111-G29M00 ISERA111-G29N00 ISERA111-G29P00 ISERA111-G29Q00 ISERA111-G29R00 ISERA111-G29S00 ISERA111-G29T00 ISERA111-G29U00 ISERA111-G29V00 ISERA111-G29W00 ISERA111-G29X00 ISERA111-G29Y00 ISERA111-G29Z00 ISERA112-G25A30 ISERA112-G25B30 ISERA112-G25C30 ISERA112-G25E30 ISERA112-G25F30 ISERA112-G25G30 ISERA112-G25A00 ISERA112-G25B00 ISERA112-G25C00 ISERA112-G25E00 ISERA112-G25F00 ISERA112-G25G00 ISERA112-G23A30 ISERA112-G23B30 ISERA112-G23C30 ISERA112-G23E30 ISERA112-G23F30 ISERA112-G23G30 ISERA112-G23A00 ISERA112-G23B00 ISERA112-G23C00 ISERA112-G23E00 ISERA112-G23F00 ISERA112-G23G00 ISERA112-G29A30 ISERA112-G29B30 ISERA112-G29C30 ISERA112-G29E30 ISERA112-G29F30 ISERA112-G29G30 ISERA112-G29A00 ISERA112-G29B00 ISERA112-G29C00 ISERA112-G29E00 ISERA112-G29F00 ISERA112-G29G00
ISAspher HPLC Columns, Diol, HILIC
Kromatografikolonner
HPLC column of the ISAspher line guarantee the highest possible reliability in a wide range of applications.
  • The production of the silica used as well as the functionalisation are carried out according to highly developed and validated process
  • Reliable and proven
  • Flexibility and robustness for every HPLC application
This ensure unchanged properties of the stationary phases in terms of selectivity and consistent chemical properties.
Direct scale-up e.g. starting from a particle size of 5 µm to semi-preparative or preparative applications (up to 45 µMore Product Information