Supelco® Ascentis® Express AQ-C18 HPLC and UHPLC Columns
Leverandør: Merck
577329-U 577321-U 565349-U 565351-U 565362-U 565363-U 565364-U 565365-U 577160-U 577320-U 577323-U 577332-U 577335-U 581363-U 582702-U 584572-U 584573-U 584574-U 584575-U 584576-U 584577-U 584578-U 584579-U 584580-U 584581-U 577161-U 577162-U 584582-U 584583-U 584584-U 577338-U 577339-U 577322-U 577324-U 577325-U 577327-U 577328-U 577330-U 577331-U 577333-U 577334-U 577336-U 577337-U 577340-U 577326-U 577341-U 584526-U 577168-U
577329-UEA 10525 NOK
577329-U 577321-U 565349-U 565351-U 565362-U 565363-U 565364-U 565365-U 577160-U 577320-U 577323-U 577332-U 577335-U 581363-U 582702-U 584572-U 584573-U 584574-U 584575-U 584576-U 584577-U 584578-U 584579-U 584580-U 584581-U 577161-U 577162-U 584582-U 584583-U 584584-U 577338-U 577339-U 577322-U 577324-U 577325-U 577327-U 577328-U 577330-U 577331-U 577333-U 577334-U 577336-U 577337-U 577340-U 577326-U 577341-U 584526-U 577168-U
Supelco® Ascentis® Express AQ-C18 HPLC and UHPLC Columns
Kromatografikolonner
Ascentis® Express AQ-C18 are high-speed, high-performance liquid chromatography columns, based on the Fused-Core® particle design.
The particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0,5 μm thick porous shell and the small overall particle size of 2,7 μm. The AQ-C18 is a C18 bonded phase prepared using a proprietary procedure that incorporates a small amount of polar silane which makes the phase resistant to dewetting. This resistance toMore Product Information
Bestill nå
SPESIFIKASJONER
Beskrivelse 2,0 µm columns 2,7 µm columns 5,0 µm columns 2,7 µm columns 2,7 µm columns 5,0 µm columns 2,5 µm columns
Environmentally Preferable
Karbonbelastning 6,5% 6,7% 5,6% 6,7% 6,5% 5,6% 6,5%
Kappet ved endene Ja Ja Ja Ja Ja Ja Ja
Maks. trykk 1000 bar 600 bar 600 bar 600 bar
pH område 2 - 9 2 - 9 2 - 9 2 - 9 2 - 9 2 - 9 2 - 9
Overflate areal 120 m²/g 135 m²/g 90 m²/g 135 m²/g 135 m²/g 90 m²/g 120 m²/g
USP kode L1 L1 L1 L1 L1 L1 L1