Pipetter, Enkanal, justerbart volum
Leverandør: EDVOTEK
589 590 591 589-1 589-2 591-1 592-1 593-1
613-1356EA 1350 NOK
613-1356 613-1359 613-1360 613-1357 613-1358 613-1361 613-1362 613-1363
Pipetter, Enkanal, justerbart volum
Pipetter Pipetter manuelle og elektroniske
Sturdily designed handheld pipettes with volumes ranging from 0,1 to 5000 µl. They are easy to use, accurate and use standard micropipette tips.
  • Volume easily selected by twisting the top
  • Lightweight design and tip ejector makes operation fast and easy
  • Tool and instructions included for self-calibration