Fuel cell car science kit
Leverandør: EDUCATIONSCIENCE COLLECTION P
765-1429EA 0 NOK
765-1429
Fuel cell car science kit
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
The Fuel Cell Car Science Kit uses a reversible PEM fuel cell that combines electrolysis and power conversion into one device. Watch as oxygen and hydrogen gases are formed in two transparent water containers. The car steers independently of the user once in operation: when the car hits a barrier, it will automatically find its way by reversing away 90 degrees. This fun science kit combines cutting-edge science, education and enjoyment.