Cars, gear drive
Leverandør: EDUCATIONSCIENCE COLLECTION P
766-0319EA 0 NOK
766-0319
Cars, gear drive
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
Car with gear drive in the series Cars and power transmission. The kit contains a corrugated plastic chassis, wheels, axles and shoulder stands with mounting pads, two gears, electric motor, battery holder and switch. Size 20×14 cm. Battery two pcs AA ordered separately.