Tires for spool wheels
Leverandør: EDUCATIONSCIENCE COLLECTION P
766-0322EA 0 NOK
766-0322
Tires for spool wheels
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
Rubber / plastic hose for the manufacture of tires for wire reels. Length 25 cm. One hose is enough for seven tires. Pack of four pcs.