Battery holder, 2xAA
Leverandør: EDUCATIONSCIENCE COLLECTION P
766-0287EA 0 NOK
766-0287
Battery holder, 2xAA
Batteriholdere
Holder for two batteries size AA. Connected in series, provides output voltage 3V. Connected with battery clips, art. No. 17-141. Pack of 5 pcs.