Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥90%
Leverandør: AAT BIOQUEST
Warning
4504 4505 4506
AATB4504EA 4500 NOK
AATB4504 AATB4505 AATB4506
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥90%
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Sulfo-SMCC is the water-soluble version of SMCC.
Formel: C₁₆H₁₈N₂O₉S
MW: 436,37 g/mol
MDL nummer: MFCD00054978
CAS-nummer: 92921-24-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fe24ecb8d003df1ec7fe314370bca9891576fa9&vid=87f000e8cab7c3fa2be70b7c58ca2a7ac0d92dde&prid=web&clid=SAPDAM