Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Alexa Fluor® 680)

Leverandør: AAT BIOQUEST
16712 16838
AATB16712EA 1800 NOK
AATB16712 AATB16838
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Alexa Fluor® 680)
Antistoffer
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

NB: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: Alexa Fluor® 680
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG
Antigen-navn Immunoglobulin G
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Klon IgG
Reaktivitet Rabbit
Vert Goat
Isotype IgG (H, L)
Kyssabsorpsjon Min×Human

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR