Ac-IETD-CHO
Leverandør: AAT Bioquest
AATB13412EA 3475 NOK
AATB13412
Ac-IETD-CHO
Proteiner
Ac-IETD-CH₂O₂ is a caspase8 inhibitor that blocks the cleavage of the 32kD caspase3 precursor into the p12 and p20subunits, thus blocking the formation of active caspase8.
  • Hazard Class/ Packing Group: 5, II
  • GHS symbol: GHS07
  • Signal Word: Warning