Native Bovine Bovine Serum Albumin (from Serum), Danofloxacin
Leverandør: AAT Bioquest
AATB50017EA 8650 NOK
AATB50017
Native Bovine Bovine Serum Albumin (from Serum), Danofloxacin
Proteiner
Danofloxacin-BSA Conjugate is used for developing anti-danofloxacin antibodies that are typically used in an ELISA assay.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
NB: For research use only.