Screen Quest™ Calbryte-520 Probenecid-Free and Wash-Free Calcium Assay Kits
Leverandør: AAT BIOQUEST
36317 36318 36319
AATB36317EA 3600 NOK
AATB36317 AATB36318 AATB36319
Screen Quest™ Calbryte-520 Probenecid-Free and Wash-Free Calcium Assay Kits
Assay Cellulær analyse
Calcium flux assays are the preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
  • 100 to 250 times fluorescence increase once bound to calcium
  • No probenecid required
  • Contains all necessary components
  • No wash step required
  • Robust and adaptable protocol
  • Brighter and more photostable signal
NB: For research use only.