Screen Quest™ Fura-2 No Wash Calcium Assay Kits
Leverandør: AAT BIOQUEST
36321 36320
AATB36321EA 53475 NOK
AATB36321 AATB36320
Screen Quest™ Fura-2 No Wash Calcium Assay Kits
Assay Cellulær analyse
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
  • Ratiometric detection
  • Contains all necessary components
  • No wash step required
  • Robust and adaptable protocol
  • Multiple research applications
NB: For research use only.