Native Amanita phalloides Phalloidin (from Amanita phalloides), Alexa Fluor® 790
Leverandør: AAT Bioquest
AATB23131EA 5175 NOK
AATB23131
Native Amanita phalloides Phalloidin (from Amanita phalloides), Alexa Fluor® 790
Proteiner
This infrared fluorescent phalloidin conjugate selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates.
  • Brighter signal
  • High affinity and selectivity
  • In house quality control
NB: For research use only.