Fluo-8FF™ potassium salt
Leverandør: AAT BIOQUEST
Warning
21102 21103
AATB21102EA 3600 NOK
AATB21102 AATB21103
Fluo-8FF™ potassium salt
Fluo-8FF™ potassium salt
Calcium measurements are critical for numerous biological investigations.
  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Cell impermeant
  • Suitable for all fluorescence platforms
NB: For research use only.
MW: 913,05 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b8ca1c7d2adf6e493367f128bf02eab61bb3ea4&vid=10720b497f99bfcd321ee5b7ff04bce8b92f46a4&prid=web&clid=SAPDAM