N-(tert-Butoxycarbonyl)-1,5-diaminopentane
Leverandør: AAT BIOQUEST
Warning
Synonymer: N-Boc-1,5-diaminopentane, tert-Butyl N-(5-Aminopentyl)carbamate, tert-Butyl (5-aminopentyl)carbamate, N-(5-Aminoamyl)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)-1,5-pentanediamine, N-(5-Aminopentyl)carbamic Acid tert-Butyl Ester, NH2-C5-NH-Boc, N-Boc-1,5-pentanediamine
4010 4011
AATB4010EA 13700 NOK
AATB4010 AATB4011
N-(tert-Butoxycarbonyl)-1,5-diaminopentane
N-(tert-Butoxycarbonyl)-1,5-diaminopentane
N-BOC-cadaverine is a monoprotected diamine that is widely used as a building block in bioconjugate chemistry.
Formel: C₁₀H₂₂N₂O₂
MW: 202,30 g/mol
MDL nummer: MFCD00210020
CAS-nummer: 51644-96-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ce7e2b09f4b90a4135607939b5fa0d69c6985d1&vid=a5a7f2b7df191c9ccf61b4754fad5069439528d6&prid=web&clid=SAPDAM