Phenylboronic acid pinacol ester
Leverandør: Apollo Scientific
Warning
Synonymer: (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, Benzeneboronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-phenyl-1,3,2-dioxaborolane, 2-Phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
OR11007-25G OR11007-100G OR11007-500G
APOSOR11007-25GEA 630 NOK
APOSOR11007-25G APOSOR11007-100G APOSOR11007-500G
Phenylboronic acid pinacol ester
Phenylboronic acid pinacol ester
Formel: C₁₂H₁₇BO₂
MW: 204,08 g/mol
Smeltepunkt: 27 - 31 °C
Flash punkt: 107 °C
MDL nummer: MFCD00966985
CAS-nummer: 24388-23-6