N-Metylantranilinsyre 99%
Leverandør: Apollo Scientific
|
Warning
|
Synonymer:
2-(Methylamino)benzoic acid
OR10023-1G
OR10023-5G
APOSOR10023-1GEA
270
NOK
APOSOR10023-1G
APOSOR10023-5G
N-Metylantranilinsyre 99%
N-Metylantranilinsyre
Formel:
C₈H₉NO₂ MW: 151,16 g/mol Smeltepunkt: 170…172 °C |
MDL nummer:
MFCD00002424 CAS-nummer: 119-68-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...