4,4'-Bis(dimetylamino)bensofenon

Leverandør: Apollo Scientific
Danger

Synonymer: 4,4′-Bis(dimethylamino)benzophenone, 4,4-Bis(dimethylamino)benzophenone, 4,4'-Di(dimethylamino)benzophenone, Michler's ketone, Bis[4-(dimethylamino)phenyl]methanone

OR72059-100G OR72059-500G
APOSOR72059-100GEA 1050 NOK
APOSOR72059-100G APOSOR72059-500G
4,4'-Bis(dimetylamino)bensofenon
4,4'-Bis(dimetylamino)bensofenon
Formel: C₁₇H₂₀N₂O
MW: 268,36 g/mol
Kokepunkt: 360 °C
Smeltepunkt: 171 - 176 °C
Flash punkt: 220 °C
MDL nummer: MFCD00008312
CAS-nummer: 90-94-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR