Bly tetraacetat 96%
Leverandør: Apollo Scientific
Warning
Synonymer: Lead(IV) tetraacetate, Triacetyloxyplumbyl acetate, Acetic acid lead (IV) salt, Lead (IV) acetate, LTA
OR924077-5G OR924077-25G OR924077-100G
APOSOR924077-5GEA 270 NOK
APOSOR924077-5G APOSOR924077-25G APOSOR924077-100G
Bly tetraacetat 96%
Bly tetraacetat
Formel: (H₃CCOO)₄Pb
MW: 443,38 g/mol
Smeltepunkt: 175 - 180 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00008693
CAS-nummer: 546-67-8
FN: 1616
ADR: 6.1,III