Experimental kit, optics
Leverandør: EDUCATIONSCIENCE COLLECTION P
765-0282EA 6600 NOK
765-0282 765-0280 765-1355
Experimental kit, optics
Lærings materiell Læremateriale for fysikk Optics Learning Activities
The kit is primarily intended for compulsory school upper secondary school or the corresponding level of knowledge. With the additional equipment, the kit is also suitable for upper secondary school level. The equipment can be used for both student labs and demonstrations. The guide describes more than fifty experiments with the presentation of theoretical foundations. No special equipment is needed other than the contents of the kit with the exception of an experiment where chemicals are required. The kit is delivered in a specially decorated wooden box. Examples of experiments.
  • Nature and distribution of light
  • Lighting and brightness
  • Reflection in flat, convex and concave mirrors
  • Breaking between different media in different symmetrical bodies etc
  • Lenses, focal length, imaging and lens defects
  • The eye and the magnifying glass
  • Camera, projector, binoculars and microscope
  • Spectrum, light scattering and colors