Demonstration instruments for measuring inserts
Leverandør: PHYWE
11100.00 11101.01 11102.01 11102.11 11102.21 11102.31 11102.33 11102.41 11102.71 11102.81 11102.83 11103.01 11103.03 11103.11 11103.21 11103.31 11103.33 11103.41 11103.81 11104.13 11104.21 11104.23 11104.31 11104.33 11104.41 11104.43 11104.51 11104.53 11105.31 11105.33 11105.41 11105.43 11105.51 11105.53 11104.65 11104.61 11102.03 11102.13 11102.23 11103.83 11103.13 11103.23
765-1247EA 0 NOK
765-1247 765-1248 765-1249 765-1251 765-1253 765-1255 765-1256 765-1257 765-1258 765-1259 765-1260 765-1261 765-1262 765-1263 765-1265 765-1267 765-1268 765-1269 765-1270 765-1272 765-1273 765-1274 765-1275 765-1276 765-1277 765-1278 765-1279 765-1280 765-1283 765-1284 765-1285 765-1286 765-1287 765-1288 765-1282 765-1281 765-1250 765-1252 765-1254 765-1271 765-1264 765-1266
Demonstration instruments for measuring inserts
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
With a special addition, the analogue instrument can be turned into a digital instrument with simultaneous analogue display. In order to extend the measurement possibilities, there are also measurements for power and voltage amplification. The replaceable measuring areas are slid into a slot on the top of the instrument ('toaster principle').
The operation can be performed by hand. Special measurements eg for Resistance and Lux is inserted horizontally from the back. All switches are done automatically via a 16-pin connection list. Most measuring ranges are passive and are thereforeMore Product Information