Precision dynamometers
Leverandør: PHYWE
03060.01 03061.01 03060.02 03060.03 03060.04 03060.06
765-1167EA 0 NOK
765-1167 765-1173 765-1168 765-1169 765-1170 765-1172
Precision dynamometers
Dynamometer
Newton double-scale dynamometers and overload protection. Zero adjustment up to 19 mm with adjusting screw for compensation of different loads eg. when tilted depending on the test setup. Skull length 12 cm for 0,1 mN and 11 cm for the other dynamometers in this series. Measuring accuracy ±0,5% of the measured value. Dynamometer cover length 18 cm.