2-Amino-5-chlorobenzoxazole 97%

Leverandør: Thermo Scientific
Danger

Synonymer: 5-Chlorobenzo[d]oxazol-2-amine, Zoxazolamine, 2-Amino-5-chloro-1,3-benzoxazole

103380050 103380250 103380010
ACRO103380050EA 935 NOK
ACRO103380050 ACRO103380250 ACRO103380010
2-Amino-5-chlorobenzoxazole 97%
2-Amino-5-chlorobenzoxazole
Formel: C₇H₅ClN₂O
MW: 168,58 g/mol
Kokepunkt: 309 °C (1013 hPa)
Smeltepunkt: 180…185 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005770
CAS-nummer: 61-80-3
EINECS: 200-519-4
FN: 2811
ADR: 6.1,III
Merck Indeks: 12,10328

Bestill nå

Spesifikasjon Testresultater

Appearance Off-white to yellow to brown Powder or crystals
Infrared spectrum Conforms
Melting point 177 to 185 °C
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR