m-Tolylacetic acid 97%
Leverandør: Thermo Scientific
Warning
Synonymer: 2-(m-Tolyl)acetic acid, (3-Methylphenyl)acetic acid
139110050 139110250 139112500
ACRO139110050EA 395 NOK
ACRO139110050 ACRO139110250 ACRO139112500
m-Tolylacetic acid 97%
m-Tolylacetic acid
Formel: (C₆H₄(CH₃))CH₂COOH
MW: 150,18 g/mol
Kokepunkt: 120 - 123 °C (26 -rr)
Smeltepunkt: 61…66 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004340
CAS-nummer: 621-36-3
EINECS: 210-683-9
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to off-white Glistening flakes and/or chunks
Infrared spectrum Conforms
Melting point 61 to 66 °C
HPLC ≥96.0 %