5-Iodouracil 99%
Leverandør: Thermo Scientific
|
Warning
|
Synonymer:
5-Iodopyrimidine-2,4(1H,3H)-dione, Iodouracil
228160100
228160500
ACRO228160100EA
1175
NOK
ACRO228160100
ACRO228160500
5-Iodouracil 99%
5-Iodouracil
Formel:
C₄H₃IN₂O₂ MW: 237,98 g/mol Kokepunkt: 401 °C (1013 hPa) Smeltepunkt: 270…274 °C (dec.) Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00006020 CAS-nummer: 696-07-1 EINECS: 211-788-2 |
Spesifikasjon Testresultater
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 266 to 276 °C (Subl) |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...