Sodium perborate monohydrate 95%

Leverandør: Thermo Fisher Scientific
Danger

386410250 386412500 386410010
ACRO386410250EA 455 NOK
ACRO386410250 ACRO386412500 ACRO386410010
Sodium perborate monohydrate 95%
Sodium perborate monohydrate
Formel: BH₂NaO₄·1H₂O
MW: 117.83 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149230
CAS-nummer: 10332-33-9
EINECS: 239-172-9
FN: 3377
ADR: 5.1 III

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Titration Iodometric ≥94 % (active oxygen)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR