N-(2-Hydroxyethyl)phthalimide ≥98%
Leverandør: Thermo Scientific
Synonymer: 2-(2-Hydroxyethyl)isoindoline-1,3-dione
A14259.22 A14259.36
A14259.22EA 740 NOK
A14259.22 A14259.36
N-(2-Hydroxyethyl)phthalimide ≥98%
N-(2-Hydroxyethyl)phthalimide
Purity: 98+% CAS No.: 3891-07-4 Molecular Formula: C10H9NO3 Formula Weight: 191.19 Melting Point: 126-129° Beilstein Reference: 147214
Formel: C₁₀H₉NO₃
MW: 191,19 g/mol
Smeltepunkt: 126…129 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005903
CAS-nummer: 3891-07-4
EINECS: 223-434-4